Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,7-Dibromo-8-hydroxyquinoline, MP Biomedicals

Catalog No. ICN211985 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN211985 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN211985 Supplier MP Biomedicals, Inc Supplier No. 0521198525
Add to Cart
Add to Cart

Chemical Identifiers

CAS 521-74-4
Molecular Formula C9H5Br2NO
Molecular Weight (g/mol) 302.953
InChI Key ZDASUJMDVPTNTF-UHFFFAOYSA-N
Synonym broxyquinoline, 5,7-dibromo-8-hydroxyquinoline, 5,7-dibromo-8-quinolinol, broxyquinolin, dibromoxin, dibromoxine, dibromoxyquinoline, broxiquinoline, dibromoquin, broxykinolin
PubChem CID 2453
IUPAC Name 5,7-dibromoquinolin-8-ol
SMILES C1=CC2=C(C(=C(C=C2Br)Br)O)N=C1

Specifications

Quantity 100 g
Formula Weight 302.96
Chemical Name or Material 5, 7-Dibromo-8-hydroxyquinoline
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.