Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,7-dichloro-[1,3]thiazolo[5,4-d]pyrimidine | 13479-88-4 | MFCD09702027 | ≥98.0% | 206.05 g/mol | C5HCl2N3S | 250 MG
SDP

Catalog No. 5000302375
Encompass_Preferred

5,7-Dichlorothiazolo[5,4-d]pyrimidine (CAS 13479-88-4) is a chlorinated heterocyclic building block used as an intermediate in small-molecule and medicinal chemistry synthesis. It is supplied in research-scale quantities and is offered by chemical suppliers in defined purity grades for use in route scouting and heterocycle construction.

  • Used as a synthetic building block for heterocycle construction and medicinal chemistry.
  • Molecular formula C5HCl2N3S and molecular weight 206.05 g/mol.
  • CAS number 13479-88-4 for unambiguous identification.
  • Offered in small pack sizes suitable for research (50 mg-500 mg).
  • Available in high-purity grades (reported ≥98% by some suppliers).
  • Supplied with standard documentation such as COA and SDS for quality verification.

Catalog No. 50-003-02375 Supplier Medchemexpress LLC Supplier No. 250MGHYW017930
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5,7-Dichlorothiazolo[5,4-d]pyrimidine (CAS 13479-88-4) is a chlorinated heterocyclic building block used as an intermediate in small-molecule and medicinal chemistry synthesis. It is supplied in research-scale quantities and is offered by chemical suppliers in defined purity grades for use in route scouting and heterocycle construction.

  • Used as a synthetic building block for heterocycle construction and medicinal chemistry.
  • Molecular formula C5HCl2N3S and molecular weight 206.05 g/mol.
  • CAS number 13479-88-4 for unambiguous identification.
  • Offered in small pack sizes suitable for research (50 mg-500 mg).
  • Available in high-purity grades (reported ≥98% by some suppliers).
  • Supplied with standard documentation such as COA and SDS for quality verification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.