Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5,7-Dichloro-1H-indole-2-carboxylic acid

Catalog No. CDS007903 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
2.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS007903 2.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS007903 Supplier Sigma Aldrich Supplier No. CDS007903250MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-butyn-2-one undergoes asymmetric double-michael reaction with ortho-tosylamidophenyl malonate catalyzed by chiral aminophosphines to yield indolines
  • It undergoes double michael reaction with nitrogen-containing tethered diacid to give pipecolic acid derivatives.3-butyn-2-one was used in the synthesis of clerodane diterpenoid (+/-)-sacacarin
  • It was used as substrate in stereoselective, conjugate arylation mediated by gallium(iii) chloride leading to (e)-α,α-unsaturated ketones

Specifications

Molecular Formula C9H5Cl2NO2
Linear Formula C9H5Cl2NO2
MDL Number MFCD02664420
Quantity 2.5 mL
Molecular Weight (g/mol) 230.05
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.