Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,7-dihydroxycoumarin | 2732-18-5 | MFCD00488354 | 99.5% | 178.14 g/mol | C9H6O4 | 250 MG
SDP

Catalog No. 5000208688
Encompass_Preferred

5,7-Dihydroxycoumarin is a research-grade coumarin isolated from the inflorescences of Macaranga triloba. It is supplied at high purity and is used in biochemical and pharmacological studies, including investigations of antibacterial and antioxidant activity.

  • High purity: 99.5% (reported)
  • Available in multiple pack sizes, including 250 mg
  • Soluble in DMSO; in vitro solubility reported at 25 mg/mL
  • Used in studies of antibacterial and antioxidant properties
  • Molecular formula C9H6O4 and molecular weight 178.14 g/mol

Catalog No. 50-002-08688 Supplier Medchemexpress LLC Supplier No. 250MGHYW072009
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5,7-Dihydroxycoumarin is a research-grade coumarin isolated from the inflorescences of Macaranga triloba. It is supplied at high purity and is used in biochemical and pharmacological studies, including investigations of antibacterial and antioxidant activity.

  • High purity: 99.5% (reported)
  • Available in multiple pack sizes, including 250 mg
  • Soluble in DMSO; in vitro solubility reported at 25 mg/mL
  • Used in studies of antibacterial and antioxidant properties
  • Molecular formula C9H6O4 and molecular weight 178.14 g/mol

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.