Learn More
Medchemexpress LLC 5,7-Dimethoxy-3-(4-pyridinyl)quinoline dihydrochloride | 1123491-15-5 | 99.1% | 339.22 | 10 MG
SDP
Catalog No. 5000345506
Encompass_Preferred
DMPQ dihydrochloride is a potent and selective inhibitor of human platelet-derived growth factor receptor β (PDGFRβ) with an IC50 of 80 nM. It is for research use only and not sold to patients.
  • Potent and selective inhibitor of human PDGFRβ.
Catalog No. 50-003-45506 Supplier Medchemexpress LLC Supplier No. HY10862710MG
DMPQ dihydrochloride is a potent and selective inhibitor of human platelet-derived growth factor receptor β (PDGFRβ) with an IC50 of 80 nM. It is for research use only and not sold to patients.
  • Potent and selective inhibitor of human PDGFRβ.