Learn More
Medchemexpress LLC 5,7-dimethoxy-3-(4-pyridinyl)quinoline dihydrochloride | 1123491-15-5 | MFCD02262123 | 99.1% | 339.22 g/mol | C16H16Cl2N2O2 | 5 MG
SDP
Catalog No. 5000263833
Encompass_Preferred
DMPQ dihydrochloride is the dihydrochloride salt of 5,7-dimethoxy-3-(4-pyridinyl)quinoline. It is a small-molecule inhibitor that selectively targets human platelet-derived growth factor receptor β (PDGFRβ) with reported biochemical potency, and is supplied as a light yellow to yellow solid for research use in biochemical and cell-based assays.
  • Potent PDGFRβ inhibition with reported IC50 of 80 nM.
  • High reported purity suitable for biochemical assays.
  • Solid form, convenient for weighing and formulation.
  • Available in multiple small pack sizes for research applications.
Catalog No. 50-002-63833 Supplier Medchemexpress LLC Supplier No. HY1086275MG
DMPQ dihydrochloride is the dihydrochloride salt of 5,7-dimethoxy-3-(4-pyridinyl)quinoline. It is a small-molecule inhibitor that selectively targets human platelet-derived growth factor receptor β (PDGFRβ) with reported biochemical potency, and is supplied as a light yellow to yellow solid for research use in biochemical and cell-based assays.
  • Potent PDGFRβ inhibition with reported IC50 of 80 nM.
  • High reported purity suitable for biochemical assays.
  • Solid form, convenient for weighing and formulation.
  • Available in multiple small pack sizes for research applications.