Learn More
Medchemexpress LLC 5,7-dimethoxy-3-pyridin-4-ylquinoline;dihydrochloride | 1123491-15-5 | MFCD02262123 | 99.1% | 339.22 | 1 MG
SDP
Catalog No. 5000346225
Encompass_Preferred
DMPQ dihydrochloride is a potent and selective inhibitor of human platelet-derived growth factor receptor β (PDGFRβ) with an IC50 of 80 nM. It is for research use only and not sold to patients.
  • Potent and selective inhibitor of human platelet-derived growth factor receptor β (PDGFRβ)
  • Inhibits PDGFRβ with an IC50 of 80 nM, useful for research applications.
Catalog No. 50-003-46225 Supplier Medchemexpress LLC Supplier No. HY10862750MG
DMPQ dihydrochloride is a potent and selective inhibitor of human platelet-derived growth factor receptor β (PDGFRβ) with an IC50 of 80 nM. It is for research use only and not sold to patients.
  • Potent and selective inhibitor of human platelet-derived growth factor receptor β (PDGFRβ)
  • Inhibits PDGFRβ with an IC50 of 80 nM, useful for research applications.