Learn More
Cayman Chemical 5,7-Dimethoxyflavone, Purity: ≥98%, CAS: 21392-57-4, 50 mg
SDP
Catalog No. 502917776
Encompass_Preferred
A flavonoid with diverse biological activities; induces the production of ROS, cell cycle arrest at the G1 phase, and apoptosis in HepG2 cells at 10, 25, and 50 µM; reduces exudate volume and inhibits prostaglandin production in a rat model of carrageenan-induced pleurisy from 75-150 mg/kg; reduces rectal temperature in a rat model of yeast-induced hyperthermia; reduces serum triglyceride, total cholesterol, and LDL levels, as well as increases serum insulin levels and reduces blood glucose levels in a rat model of STZ-induced diabetes
Catalog No. 50-291-7776 Supplier Cayman Chemical Supplier No. 3736150 MG
A flavonoid with diverse biological activities; induces the production of ROS, cell cycle arrest at the G1 phase, and apoptosis in HepG2 cells at 10, 25, and 50 µM; reduces exudate volume and inhibits prostaglandin production in a rat model of carrageenan-induced pleurisy from 75-150 mg/kg; reduces rectal temperature in a rat model of yeast-induced hyperthermia; reduces serum triglyceride, total cholesterol, and LDL levels, as well as increases serum insulin levels and reduces blood glucose levels in a rat model of STZ-induced diabetes