Learn More
Medchemexpress LLC 5,7-Dimethoxyflavone | 21392-57-4 | 1 ML
SDP
Catalog No. 5000418658
Encompass_Preferred
5,7-Dimethoxyflavone is a major component of *Kaempferia parviflora* with diverse biological activities. It exhibits anti-obesity, anti-inflammatory, and antineoplastic effects. This compound acts as an inhibitor of cytochrome P450 (CYP) 3As and is a potent Breast Cancer Resistance Protein (BCRP) inhibitor.
  • Exhibits anti-obesity, anti-inflammatory, and antineoplastic effects
  • Acts as a potent inhibitor of CYP3A and BCRP
  • Inhibits Bcrp1-mediated Sorafenib efflux
  • Induces melanogenesis
  • Inhibits lipid accumulation and adipocyte differentiation
  • Reduces liver CYP3A11 and CYP3A25 protein expression
  • Inhibits sarcopenia
  • Decreases body weight gain and inhibits fatty liver
Catalog No. 50-004-18658 Supplier Medchemexpress LLC Supplier No. HYN501110MM/1ML
5,7-Dimethoxyflavone is a major component of *Kaempferia parviflora* with diverse biological activities. It exhibits anti-obesity, anti-inflammatory, and antineoplastic effects. This compound acts as an inhibitor of cytochrome P450 (CYP) 3As and is a potent Breast Cancer Resistance Protein (BCRP) inhibitor.
  • Exhibits anti-obesity, anti-inflammatory, and antineoplastic effects
  • Acts as a potent inhibitor of CYP3A and BCRP
  • Inhibits Bcrp1-mediated Sorafenib efflux
  • Induces melanogenesis
  • Inhibits lipid accumulation and adipocyte differentiation
  • Reduces liver CYP3A11 and CYP3A25 protein expression
  • Inhibits sarcopenia
  • Decreases body weight gain and inhibits fatty liver