Learn More
Medchemexpress LLC 5,7-Dimethoxyflavone | 21392-57-4 | 100 MG
SDP
Catalog No. 5000418034
Encompass_Preferred
5,7-Dimethoxyflavone is a natural compound found in *Kaempferia parviflora* that exhibits diverse biological activities. It possesses anti-obesity, anti-inflammatory, and antineoplastic effects. This compound is known to inhibit cytochrome P450 (CYP) 3As and acts as a potent Breast Cancer Resistance Protein (BCRP) inhibitor.
  • Inhibits cytochrome P450 (CYP) 3As
  • Potent Breast Cancer Resistance Protein (BCRP) inhibitor
  • Exhibits anti-obesity effects
  • Demonstrates anti-inflammatory properties
  • Shows antineoplastic activity
  • Reduces liver CYP3A11 and CYP3A25 protein expression in mice
  • Inhibits sarcopenia in aged mice
  • Decreases body weight gain and inhibits fatty liver in HFD mice
Catalog No. 50-004-18034 Supplier Medchemexpress LLC Supplier No. HYN5011100MG
5,7-Dimethoxyflavone is a natural compound found in *Kaempferia parviflora* that exhibits diverse biological activities. It possesses anti-obesity, anti-inflammatory, and antineoplastic effects. This compound is known to inhibit cytochrome P450 (CYP) 3As and acts as a potent Breast Cancer Resistance Protein (BCRP) inhibitor.
  • Inhibits cytochrome P450 (CYP) 3As
  • Potent Breast Cancer Resistance Protein (BCRP) inhibitor
  • Exhibits anti-obesity effects
  • Demonstrates anti-inflammatory properties
  • Shows antineoplastic activity
  • Reduces liver CYP3A11 and CYP3A25 protein expression in mice
  • Inhibits sarcopenia in aged mice
  • Decreases body weight gain and inhibits fatty liver in HFD mice