Learn More
Medchemexpress LLC 5,7-Dimethoxyflavone | 21392-57-4 | 50 MG
SDP
Catalog No. 5000418035
Encompass_Preferred
5,7-Dimethoxyflavone is an analytical standard and a major component of Kaempferia parviflora. It exhibits anti-obesity, anti-inflammatory, and antineoplastic effects. This product is intended for research and analytical applications.
  • Inhibits cytochrome P450 (CYP) 3As
  • Potent breast cancer resistance protein (BCRP) inhibitor
  • Used for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Shipping is at room temperature in continental US; may vary elsewhere. Store under recommended conditions in the Certificate of Analysis.
Catalog No. 50-004-18035 Supplier Medchemexpress LLC Supplier No. HYN5011R50MG
5,7-Dimethoxyflavone is an analytical standard and a major component of Kaempferia parviflora. It exhibits anti-obesity, anti-inflammatory, and antineoplastic effects. This product is intended for research and analytical applications.
  • Inhibits cytochrome P450 (CYP) 3As
  • Potent breast cancer resistance protein (BCRP) inhibitor
  • Used for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Shipping is at room temperature in continental US; may vary elsewhere. Store under recommended conditions in the Certificate of Analysis.