Learn More
Medchemexpress LLC 5,7-dimethoxyflavone | 21392-57-4 | MFCD00016943 | 282.29 g/mol | C17H14O4 | 10 MG
SDP
Catalog No. 5000289700
Encompass_Preferred
5,7-Dimethoxyflavone is a flavone derivative supplied as an analytical standard for research and analytical applications. It occurs naturally in Kaempferia parviflora and is used in studies of metabolic enzyme interactions, transporter inhibition, and biological activity profiling.
  • Analytical standard suitable for assay and quantification.
  • Natural flavone component used in phytochemical research.
  • Documented inhibitor activity against cytochrome P450 enzymes and BCRP, useful in metabolism and transporter studies.
  • Supplied in small mass quantities appropriate for analytical work.
  • Molecular formula C17H14O4 and molecular weight 282.29 g/mol.
Catalog No. 50-002-89700 Supplier Medchemexpress LLC Supplier No. HYN5011R10MG
5,7-Dimethoxyflavone is a flavone derivative supplied as an analytical standard for research and analytical applications. It occurs naturally in Kaempferia parviflora and is used in studies of metabolic enzyme interactions, transporter inhibition, and biological activity profiling.
  • Analytical standard suitable for assay and quantification.
  • Natural flavone component used in phytochemical research.
  • Documented inhibitor activity against cytochrome P450 enzymes and BCRP, useful in metabolism and transporter studies.
  • Supplied in small mass quantities appropriate for analytical work.
  • Molecular formula C17H14O4 and molecular weight 282.29 g/mol.