Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,7-dimethoxyflavone | 21392-57-4 | MFCD00016943 | 282.29 g/mol | C17H14O4 | 10 MG
SDP

Catalog No. 5000289708
Encompass_Preferred

5,7-Dimethoxyflavone is a dimethoxylated flavone found in Kaempferia parviflora. It is supplied as a research and analytical reagent for laboratory use and serves as an analytical standard and reference material. Reported activities include inhibition of cytochrome P450 3A enzymes and breast cancer resistance protein (BCRP).

  • CAS number: 21392-57-4.
  • Molecular formula: C17H14O4.
  • Molecular weight: 282.29 g/mol.
  • Supplied as a small solid quantity suitable for analytical testing (10 MG).
  • Intended for research and analytical use only; not for human use.
  • Store protected from light; solid storage at 4°C; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-89708 Supplier Medchemexpress LLC Supplier No. HYN501110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5,7-Dimethoxyflavone is a dimethoxylated flavone found in Kaempferia parviflora. It is supplied as a research and analytical reagent for laboratory use and serves as an analytical standard and reference material. Reported activities include inhibition of cytochrome P450 3A enzymes and breast cancer resistance protein (BCRP).

  • CAS number: 21392-57-4.
  • Molecular formula: C17H14O4.
  • Molecular weight: 282.29 g/mol.
  • Supplied as a small solid quantity suitable for analytical testing (10 MG).
  • Intended for research and analytical use only; not for human use.
  • Store protected from light; solid storage at 4°C; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.