Learn More
Medchemexpress LLC 5,7-Dimethoxyflavone | 21392-57-4 | MFCD00016943 | 5 MG
SDP
Catalog No. 5000419778
Encompass_Preferred
5,7-Dimethoxyflavone is a major component of Kaempferia parviflora. It exhibits anti-obesity, anti-inflammatory, and antineoplastic effects. It is a potent Breast Cancer Resistance Protein (BCRP) inhibitor and inhibits cytochrome P450 (CYP) 3As.
  • Exhibits anti-obesity properties
  • Demonstrates anti-inflammatory effects
  • Shows antineoplastic properties
  • Inhibits cytochrome P450 (CYP) 3As
  • Acts as a potent Breast Cancer Resistance Protein (BCRP) inhibitor
  • Used in metabolic disease research
  • Used in inflammation/immunology research
  • Used in cancer research
Catalog No. 50-004-19778 Supplier Medchemexpress LLC Supplier No. HYN5011R5MG
5,7-Dimethoxyflavone is a major component of Kaempferia parviflora. It exhibits anti-obesity, anti-inflammatory, and antineoplastic effects. It is a potent Breast Cancer Resistance Protein (BCRP) inhibitor and inhibits cytochrome P450 (CYP) 3As.
  • Exhibits anti-obesity properties
  • Demonstrates anti-inflammatory effects
  • Shows antineoplastic properties
  • Inhibits cytochrome P450 (CYP) 3As
  • Acts as a potent Breast Cancer Resistance Protein (BCRP) inhibitor
  • Used in metabolic disease research
  • Used in inflammation/immunology research
  • Used in cancer research