Learn More
Medchemexpress LLC 5,7-Dimethoxyflavone (Standard) | 21392-57-4 | 25 MG
SDP
Catalog No. 5000420555
Encompass_Preferred
5,7-Dimethoxyflavone (Standard) is the analytical standard of 5,7-Dimethoxyflavone, intended for research and analytical applications. It is a major component of Kaempferia parviflora, known for its anti-obesity, anti-inflammatory, and antineoplastic effects. This compound inhibits cytochrome P450 (CYP) 3As and is also a potent Breast Cancer Resistance Protein (BCRP) inhibitor.
  • Analytical standard for research and analytical applications
  • Major component of Kaempferia parviflora
  • Exhibits anti-obesity effects
  • Exhibits anti-inflammatory effects
  • Exhibits antineoplastic effects
  • Inhibits cytochrome P450 (CYP) 3As
  • Potent Breast Cancer Resistance Protein (BCRP) inhibitor
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
Catalog No. 50-004-20555 Supplier Medchemexpress LLC Supplier No. 25MGHYN5011R
5,7-Dimethoxyflavone (Standard) is the analytical standard of 5,7-Dimethoxyflavone, intended for research and analytical applications. It is a major component of Kaempferia parviflora, known for its anti-obesity, anti-inflammatory, and antineoplastic effects. This compound inhibits cytochrome P450 (CYP) 3As and is also a potent Breast Cancer Resistance Protein (BCRP) inhibitor.
  • Analytical standard for research and analytical applications
  • Major component of Kaempferia parviflora
  • Exhibits anti-obesity effects
  • Exhibits anti-inflammatory effects
  • Exhibits antineoplastic effects
  • Inhibits cytochrome P450 (CYP) 3As
  • Potent Breast Cancer Resistance Protein (BCRP) inhibitor
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS