Learn More
Cayman Chemical (±)-5,7-Dimethyltocol, Purity: ≥95%, CAS: 493-35-6, 50 mg
SDP
Catalog No. 502939287
Encompass_Preferred
A form of tocopherol; has similar antioxidant activity to α-tocol, but lower activity than γ-tocol; increases microviscosity of PC rat liver liposomes by 70.6% when used at a molar ratio of 0.2 to PC; has been used as an internal standard for the quantification of tocols, tocopherols, and tocopheryl quinones by HPLC
Catalog No. 50-293-9287 Supplier Cayman Chemical Supplier No. 2719450 MG
A form of tocopherol; has similar antioxidant activity to α-tocol, but lower activity than γ-tocol; increases microviscosity of PC rat liver liposomes by 70.6% when used at a molar ratio of 0.2 to PC; has been used as an internal standard for the quantification of tocols, tocopherols, and tocopheryl quinones by HPLC