Learn More
Selleck Chemical LLC 5.7-Dimethoxyflavone S3227-1mg
SDP
Catalog No. 503109104
Encompass_Preferred
5 7-Dimethoxyflavone (5 7-DMF) is an inhibitor of cytochrome P450 (CYP) 3As that markedly decreases the expression of CYP3A11 and CYP3A25 in the liver 5 7-Dimethoxyflavone (5 7-DMF) is also a potent Breast Cancer Resistance Protein (BCRP ABCG2) inhibitor 5 7-Dimethoxyflavone (5 7-DMF) is one of the major components of Kaempferia parviflora with anti-obesity anti-inflammatory and antineoplastic effects
Catalog No. 50-310-9104 Supplier Selleck Chemical LLC Supplier No. S32271MG
5 7-Dimethoxyflavone (5 7-DMF) is an inhibitor of cytochrome P450 (CYP) 3As that markedly decreases the expression of CYP3A11 and CYP3A25 in the liver 5 7-Dimethoxyflavone (5 7-DMF) is also a potent Breast Cancer Resistance Protein (BCRP ABCG2) inhibitor 5 7-Dimethoxyflavone (5 7-DMF) is one of the major components of Kaempferia parviflora with anti-obesity anti-inflammatory and antineoplastic effects