Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,7-Dodecadiyne-1,12-diol, 96%, Thermo Scientific™

Catalog No. AAL1148506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1148506 5 g
AAL1148503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1148506 Supplier Thermo Scientific Chemicals Supplier No. L1148506

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74602-32-7
Molecular Formula C12H18O2
Molecular Weight (g/mol) 194.27
MDL Number MFCD00078343
InChI Key SEWYHOMCDKWYEF-UHFFFAOYSA-N
Synonym 5,7-dodecadiyne-1,12-diol, 5,7-dodecadiyn-1,12-diol, 5,7-dodecadiyne-1,12-diol #, 5,7-dodecadiyn-1,12-diol,
PubChem CID 560878
IUPAC Name dodeca-5,7-diyne-1,12-diol
SMILES OCCCCC#CC#CCCCCO

Specifications

Melting Point 49°C to 51°C
Quantity 5 g
Beilstein 1757614
Formula Weight 194.28
Percent Purity 96%
Chemical Name or Material 5,7-Dodecadiyne-1,12-diol
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.