Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 5.75 inch Pasteur Pipets, Disposable, Bulk Pack, Non-Sterile, Unplugged

Catalog No. 08757320 Shop All Corning Products
Change view
Click to view available options
Length (English):
5.75 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
08-757-320 5.75 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-757-320 Supplier Corning™ Supplier No. 7095D5X
  • PRODUCT PROMOTION
    $345.50
    $138.00 / Case of 1000

    Save $207.50

    60% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Feature a constriction one inch below the top to facilitate plugging. Corning™ Disposable Glass Pasteur Pipets are ideal for nonvolumetric transfer work in bacteriology, immunology, hematology and serology studies.

Corning disposable glass Pasteur pipets are available in convenient Packages. One-time use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents. These pipets feature a constriction one inch below the top to facilitate plugging. They are ideal for rapid non­volumetric transfer work in bacteriology, immunology, hematology, and serology studies, as well as blood bank and clinical chemistry procedures.



'B' series pipets are soda lime glass. 'D' series are borosilicate glass.
TRUSTED_SUSTAINABILITY

Specifications

Material Borosilicate Glass
Length (English) 5.75 in.
Type Pasteur Pipet
Disposable Single-use
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.