Learn More
CELLTREAT Scientific Products 5.75 Inch Polypropylene Plasteur® Pasteur Pipet, Bulk Packed in Tab Lock Box, Non-sterile pipette
SDP
Catalog No. 501149113
Encompass_Preferred
CELLTREAT Scientific Products 5.75"""" Polypropylene Plasteur Pipet. The Plasteur Plastic Pasteur Pipet offers the aspirating precision of a glass pipet and the safety benefits of plastic. Fine tip is ideal for precision aspirating. Polypropylene provides the chemical resistivity needed for many applications. No more broken glass: more durable than standard glass pipets. Flexible plastic helps prevent glove or skin punctures. Will not shatter, eliminating losses due to broken glass. No damage during shipping and handling as compared to traditional glass pipets. Eliminate """"sharps"""" disposal. Can be autoclaved. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 200 bulk packed/tab lock box, 1000/case.
Catalog No. 50-114-9113 Supplier CELLTREAT Scientific Products Supplier No. 229274
CELLTREAT Scientific Products 5.75"""" Polypropylene Plasteur Pipet. The Plasteur Plastic Pasteur Pipet offers the aspirating precision of a glass pipet and the safety benefits of plastic. Fine tip is ideal for precision aspirating. Polypropylene provides the chemical resistivity needed for many applications. No more broken glass: more durable than standard glass pipets. Flexible plastic helps prevent glove or skin punctures. Will not shatter, eliminating losses due to broken glass. No damage during shipping and handling as compared to traditional glass pipets. Eliminate """"sharps"""" disposal. Can be autoclaved. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 200 bulk packed/tab lock box, 1000/case.