Learn More
Cayman Chemical 5,7-Dichlorothiazolo[5,4-d]pyrimidine, Purity: ≥95%, CAS: 13479-88-4, 100 mg
SDP
Catalog No. 502912116
Encompass_Preferred
A building block; has been used in the synthesis Syk, mTOR, and PI3Kδ inhibitors, as well as HIV-1 NNRTIs with antiviral activity
Catalog No. 50-291-2116 Supplier Cayman Chemical Supplier No. 36489100 MG
A building block; has been used in the synthesis Syk, mTOR, and PI3Kδ inhibitors, as well as HIV-1 NNRTIs with antiviral activity