Learn More
Cayman Chemical 5,7-Dihydroxy-4-methylcoumarin, Purity: ≥98%, CAS: 2107-76-8, 250 mg
SDP
Catalog No. 502947666
Encompass_Preferred
A synthetic coumarin with diverse biological activities; scavenges alkylperoxy radicals and HOCl (IC50 = 2.85 µM) in cell-free assays; inhibits lipid peroxidation in rat liver microsomes, A23187-induced TXB2 generation in isolated rat peritoneal leukocytes, and MPO activity in a cell-free assay (IC50s = 12, 1, and 1.06 µM, respectively); inhibits rat lens aldose reductase (IC50 = 17 µM); inhibits human CAI, CAIX, and CAXII (Kis = 8.4, 0.19, and 6.4 µM, respectively); inhibits arachidonic acid-induced platelet aggregation (IC50 = 45 µM); inhibits HCV NS5B polymerase (IC50 = 47.2 µM); active against H. pylori (MIC = 10 µg/ml)
Catalog No. 50-294-7666 Supplier Cayman Chemical Supplier No. 33756250 MG
A synthetic coumarin with diverse biological activities; scavenges alkylperoxy radicals and HOCl (IC50 = 2.85 µM) in cell-free assays; inhibits lipid peroxidation in rat liver microsomes, A23187-induced TXB2 generation in isolated rat peritoneal leukocytes, and MPO activity in a cell-free assay (IC50s = 12, 1, and 1.06 µM, respectively); inhibits rat lens aldose reductase (IC50 = 17 µM); inhibits human CAI, CAIX, and CAXII (Kis = 8.4, 0.19, and 6.4 µM, respectively); inhibits arachidonic acid-induced platelet aggregation (IC50 = 45 µM); inhibits HCV NS5B polymerase (IC50 = 47.2 µM); active against H. pylori (MIC = 10 µg/ml)