Learn More
Medchemexpress LLC 5,8,11,14,17,20-Hexaoxa-2-azadocosanedioic acid, 1-(9H-fluoren-9-ylmethyl) ester | 437655-96-4 | 99.4% | 1 G

Supplier: Medchemexpress LLC HY1303641G
Fmoc-NH-PEG6-CH2COOH is a cleavable linker used in the synthesis of antibody-drug conjugates (ADCs). It also functions as a PEG-based PROTAC linker, applicable in the synthesis of PROTACs. This compound facilitates the controlled degradation of target proteins by exploiting the intracellular ubiquitin-proteasome system.
- Cleavable linker for ADC synthesis
- PEG-based PROTAC linker
- Utilizes ubiquitin-proteasome system
- Enables selective degradation of target proteins
- Molecular weight of 561.62
- Formula C29H39NO10
- Appearance: colorless to light yellow liquid
- Soluble in DMSO (100 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.