Learn More
Medchemexpress LLC 5,8,11,14,17,20-hexaoxa-2-azadocosanedioic acid, 1-(9H-fluoren-9-ylmethyl) ester | 437655-96-4 | 99.4% | 561.62 | C29H39NO10 | 250 MG
SDP
Catalog No. 5000203236
Encompass_Preferred
Fmoc-NH-PEG6-CH2COOH is an Fmoc-protected polyethylene glycol (PEG6) linker with a terminal carboxylic acid, supplied for research use as a cleavable linker for antibody-drug conjugates and as a PEG-based PROTAC linker in targeted protein degradation research.
  • Fmoc-protected amine with terminal carboxylic acid functionality.
  • Suitable for ADC and PROTAC linker synthesis.
  • Reported purity 99.4% and molecular weight 561.62.
  • Molecular formula C29H39NO10.
  • Available in small-scale pack sizes including 250 MG.
Catalog No. 50-002-03236 Supplier Medchemexpress LLC Supplier No. HY130364250MG
Fmoc-NH-PEG6-CH2COOH is an Fmoc-protected polyethylene glycol (PEG6) linker with a terminal carboxylic acid, supplied for research use as a cleavable linker for antibody-drug conjugates and as a PEG-based PROTAC linker in targeted protein degradation research.
  • Fmoc-protected amine with terminal carboxylic acid functionality.
  • Suitable for ADC and PROTAC linker synthesis.
  • Reported purity 99.4% and molecular weight 561.62.
  • Molecular formula C29H39NO10.
  • Available in small-scale pack sizes including 250 MG.