Learn More
Medchemexpress LLC 5,8,11,14,17,20-hexaoxa-2-azatricosanedioic acid, 1-(1,1-dimethylethyl) ester | 882847-13-4 | >97.0% | 453.52 g/mol | C20H39NO10 | 250 MG
SDP
Catalog No. 5000203066
Encompass_Preferred
Boc-NH-PEG6-CH2CH2COOH is a Boc-protected PEG6 linker with a terminal carboxylic acid, supplied for use in synthetic and bioconjugation applications such as PROTAC and ADC linker assembly.
  • Boc-protected amine functionality allows orthogonal deprotection
  • Peg6 spacer improves solubility and provides flexibility
  • Terminal carboxylic acid enables amide coupling chemistry
  • High purity (≥97.0%) as determined by NMR
  • Available in small research pack sizes (250 MG)
Catalog No. 50-002-03066 Supplier Medchemexpress LLC Supplier No. HYW040244250MG
Boc-NH-PEG6-CH2CH2COOH is a Boc-protected PEG6 linker with a terminal carboxylic acid, supplied for use in synthetic and bioconjugation applications such as PROTAC and ADC linker assembly.
  • Boc-protected amine functionality allows orthogonal deprotection
  • Peg6 spacer improves solubility and provides flexibility
  • Terminal carboxylic acid enables amide coupling chemistry
  • High purity (≥97.0%) as determined by NMR
  • Available in small research pack sizes (250 MG)