Learn More
Medchemexpress LLC 5,8,11,14,17-Pentaoxa-2-azaeicosanedioic acid, 1-(1,1-dimethylethyl) 20-(2,5-dioxo-1-pyrrolidinyl) ester | 2055040-78-1 | 96.9% | 506.54 | 1 G
SDP
Catalog No. 5000438068
Encompass_Preferred
Boc-N-PEG5-C2-NHS ester is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This compound facilitates the targeted degradation of proteins by exploiting the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-004-38068 Supplier Medchemexpress LLC Supplier No. HY1263811G
Boc-N-PEG5-C2-NHS ester is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This compound facilitates the targeted degradation of proteins by exploiting the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins