Learn More
Medchemexpress LLC 5,8,11,14,17-pentaoxa-2-azaeicosanedioic acid, 1-(1,1-dimethylethyl) 20-(2,5-dioxo-1-pyrrolidinyl) ester | 2055040-78-1 | 96.9% | 506.54 | 100 MG
SDP
Catalog No. 5000436254
Encompass_Preferred
Boc-N-PEG5-C2-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker; one for an E3 ubiquitin ligase and the other for a specific target protein. These molecules function by harnessing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-004-36254 Supplier Medchemexpress LLC Supplier No. HY126381100MG
Boc-N-PEG5-C2-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker; one for an E3 ubiquitin ligase and the other for a specific target protein. These molecules function by harnessing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins