Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,8,11,14,20,23,26,29-Octaoxa-2,17,32-triazatritriacontanedioic acid, 1,33-bis(1,1-dimethylethyl) | 2182601-75-6 | ≥97.0% | C₃₀H₆₁N₃O₁₂ | 250 MG
SDP

Catalog No. 5000352876
Encompass_Preferred

NH-bis(PEG4-C2-NH-Boc) is a chemical compound, also known by its chemical name 5,8,11,14,20,23,26,29-Octaoxa-2,17,32-triazatritriacontanedioic acid, 1,33-bis(1,1-dimethylethyl) ester. It is identified by CAS number 2182601-75-6 and has a molecular formula of C₃₀H₆₁N₃O₁₂. This product appears as a colorless to light yellow liquid with a purity of ≥97.0%. It is intended for research use only and has not been fully validated for medical applications.

  • Identified by CAS number 2182601-75-6
  • Molecular formula: C₃₀H₆₁N₃O₁₂
  • Molecular weight: 655.82
  • Purity: ≥97.0%
  • Appearance: Colorless to light yellow liquid
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • For research use only

Catalog No. 50-003-52876 Supplier Medchemexpress LLC Supplier No. 250MGHY140261
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NH-bis(PEG4-C2-NH-Boc) is a chemical compound, also known by its chemical name 5,8,11,14,20,23,26,29-Octaoxa-2,17,32-triazatritriacontanedioic acid, 1,33-bis(1,1-dimethylethyl) ester. It is identified by CAS number 2182601-75-6 and has a molecular formula of C₃₀H₆₁N₃O₁₂. This product appears as a colorless to light yellow liquid with a purity of ≥97.0%. It is intended for research use only and has not been fully validated for medical applications.

  • Identified by CAS number 2182601-75-6
  • Molecular formula: C₃₀H₆₁N₃O₁₂
  • Molecular weight: 655.82
  • Purity: ≥97.0%
  • Appearance: Colorless to light yellow liquid
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.