Learn More
Medchemexpress LLC 5,8,11,14-Eicosatetraenoic-5,6,8,9,11,12,14,15-d8 acid, (5Z,8Z,11Z,14Z)- | 69254-37-1 | M002808 | 99.5% | 312.52 | 1 MG
SDP
Catalog No. 5000345141
Encompass_Preferred
Arachidonic acid-d8 is the deuterium labeled Arachidonic acid. Arachidonic acid is an essential fatty acid and a major constituent of biomembranes.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-45141 Supplier Medchemexpress LLC Supplier No. HY109590S1MG
Arachidonic acid-d8 is the deuterium labeled Arachidonic acid. Arachidonic acid is an essential fatty acid and a major constituent of biomembranes.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.