Learn More
Medchemexpress LLC 5,8,11,14-tetraoxa-2-azahexadecanedioic acid, 1-(1,1-dimethylethyl) ester | 876345-13-0 | MFCD30528568 | 97.0% | 351.39 | C15H29NO8 | 500 MG
SDP
Catalog No. 5000202207
Encompass_Preferred
Boc-NH-PEG4-CH2COOH is a Boc-protected PEG4 linker with a terminal carboxylic acid, intended for use as a cleavable linker in antibody-drug conjugate (ADC) synthesis and as a PEG-based PROTAC linker in medicinal chemistry and bioconjugation research.
  • Boc-protected amino group for orthogonal protection
  • Four-unit polyethylene glycol spacer for improved solubility and flexibility
  • Terminal carboxylic acid for amide coupling or conjugation
  • High purity suitable for research applications
  • Stable under recommended storage conditions
Catalog No. 50-002-02207 Supplier Medchemexpress LLC Supplier No. HY42640500MG
Boc-NH-PEG4-CH2COOH is a Boc-protected PEG4 linker with a terminal carboxylic acid, intended for use as a cleavable linker in antibody-drug conjugate (ADC) synthesis and as a PEG-based PROTAC linker in medicinal chemistry and bioconjugation research.
  • Boc-protected amino group for orthogonal protection
  • Four-unit polyethylene glycol spacer for improved solubility and flexibility
  • Terminal carboxylic acid for amide coupling or conjugation
  • High purity suitable for research applications
  • Stable under recommended storage conditions