Learn More
Medchemexpress LLC 5,8,11,14-tetraoxa-2-azahexadecanoic acid, 16-amino-, 1,1-dimethylethyl ester | 811442-84-9 | ≥98.0% | 336.42 g/mol | C15H32N2O6 | 25 G
SDP
Catalog No. 5000203137
Encompass_Preferred
Boc-protected PEG4 diamine used as a cleavable PEG spacer and linker in antibody-drug conjugate and PROTAC synthesis. It provides a terminal primary amine for conjugation and a Boc-protected site for selective deprotection.
  • Boc-protected terminal amine for selective deprotection.
  • PEG4 spacer for improved solubility and flexibility.
  • Suitable for ADC and PROTAC linker synthesis.
  • High purity (≥98.0% by NMR).
  • Monodisperse PEG chain with predictable molecular weight.
Catalog No. 50-002-03137 Supplier Medchemexpress LLC Supplier No. HYW00835225G
Boc-protected PEG4 diamine used as a cleavable PEG spacer and linker in antibody-drug conjugate and PROTAC synthesis. It provides a terminal primary amine for conjugation and a Boc-protected site for selective deprotection.
  • Boc-protected terminal amine for selective deprotection.
  • PEG4 spacer for improved solubility and flexibility.
  • Suitable for ADC and PROTAC linker synthesis.
  • High purity (≥98.0% by NMR).
  • Monodisperse PEG chain with predictable molecular weight.