Learn More
Medchemexpress LLC 5,8,11,14-tetraoxa-2-azahexadecanoic acid, 16-amino-, 1,1-dimethylethyl ester | 811442-84-9 | MFCD16619339 | ≥98.0% | 336.42 g/mol | C15H32N2O6 | 500 MG
SDP
Catalog No. 5000204094
Encompass_Preferred
Boc-NH-PEG4-CH2CH2NH2 (5,8,11,14-tetraoxa-2-azahexadecanoic acid, 16-amino-, 1,1-dimethylethyl ester) is a cleavable 5-unit polyethylene glycol (PEG) linker used for conjugation chemistry in ADC and PROTAC synthesis. It provides orthogonal protection with a Boc-protected amine and a free terminal amine for stepwise coupling.
  • Cleavable 5-unit PEG linker suitable for conjugation chemistry.
  • Boc-protected amine and free terminal amine for orthogonal conjugation.
  • High purity ≥98.0% for reliable reactions.
  • Molecular weight 336.42 g/mol and formula C15H32N2O6.
  • Available as a 500 MG research-use package.
Catalog No. 50-002-04094 Supplier Medchemexpress LLC Supplier No. HYW008352500MG
Boc-NH-PEG4-CH2CH2NH2 (5,8,11,14-tetraoxa-2-azahexadecanoic acid, 16-amino-, 1,1-dimethylethyl ester) is a cleavable 5-unit polyethylene glycol (PEG) linker used for conjugation chemistry in ADC and PROTAC synthesis. It provides orthogonal protection with a Boc-protected amine and a free terminal amine for stepwise coupling.
  • Cleavable 5-unit PEG linker suitable for conjugation chemistry.
  • Boc-protected amine and free terminal amine for orthogonal conjugation.
  • High purity ≥98.0% for reliable reactions.
  • Molecular weight 336.42 g/mol and formula C15H32N2O6.
  • Available as a 500 MG research-use package.