Learn More
Medchemexpress LLC 5,8,11-Trioxa-2,14-diazahexadecanoic acid, 16-bromo-15-oxo-, 1,1-dimethylethyl ester | 1421933-39-2 | ≥98.0% | C15H29BrN2O6 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY141200500MG
Encompass_Preferred
Bromoacetamido-PEG3-NH-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are designed to selectively degrade target proteins by utilizing the ubiquitin-proteasome system, connecting ligands for an E3 ubiquitin ligase and the target protein via a linker.
  • Suitable for PROTAC synthesis
  • Utilizes PEG-based linker technology
  • Facilitates targeted protein degradation
Catalog No. 50-003-55300