Learn More
Medchemexpress LLC 5,8,11-Trioxa-2-azatetradecanedioic acid, 1-(4-cycloocten-1-yl) ester | 2141981-86-2 | MFCD31811538 | 96.0% | 373.44 | 25 MG
SDP
Catalog No. 5000348730
Encompass_Preferred
TCO-PEG3-acid is a click chemistry reagent that contains a TCO group, which can undergo an inverse electron demand Diels-Alder reaction (iEDDA) with molecules containing Tetrazine groups. It is also a click chemistry PEG linker with a terminal carboxylic acid group. The terminal carboxylic acid is reactive with primary amine groups in the presence of activators to form a stable amide bond.
  • Contains a TCO group for inverse electron demand Diels-Alder reaction (iEDDA)
  • Reacts with molecules containing Tetrazine groups
  • Functions as a click chemistry PEG linker
  • Terminal carboxylic acid group
  • Forms stable amide bonds with primary amine groups in the presence of activators
Catalog No. 50-003-48730 Supplier Medchemexpress LLC Supplier No. 25MGHY140015
TCO-PEG3-acid is a click chemistry reagent that contains a TCO group, which can undergo an inverse electron demand Diels-Alder reaction (iEDDA) with molecules containing Tetrazine groups. It is also a click chemistry PEG linker with a terminal carboxylic acid group. The terminal carboxylic acid is reactive with primary amine groups in the presence of activators to form a stable amide bond.
  • Contains a TCO group for inverse electron demand Diels-Alder reaction (iEDDA)
  • Reacts with molecules containing Tetrazine groups
  • Functions as a click chemistry PEG linker
  • Terminal carboxylic acid group
  • Forms stable amide bonds with primary amine groups in the presence of activators