Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,8,11-trioxa-2-azatetradecanedioic acid, 1-(9H-fluoren-9-ylmethyl) ester | 867062-95-1 | 99.8% | 443.49 g/mol | C24H29NO7 | 1 G
SDP

Catalog No. 5000203087
Encompass_Preferred

Fmoc-NH-PEG3-CH2CH2COOH is an Fmoc-protected PEG3 linker bearing a terminal carboxylic acid, used in research applications as a cleavable linker for antibody-drug conjugates and as a PEG-based linker for PROTAC synthesis.

  • Fmoc-protected amine functionality for standard peptide chemistry
  • Terminal carboxylic acid for conjugation
  • High reported purity (≈99.8%)
  • Molecular weight 443.49 g/mol
  • Chemical formula C24H29NO7
  • Available in small-scale quantities, including 1 G

Catalog No. 50-002-03087 Supplier Medchemexpress LLC Supplier No. HYW0402311G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-NH-PEG3-CH2CH2COOH is an Fmoc-protected PEG3 linker bearing a terminal carboxylic acid, used in research applications as a cleavable linker for antibody-drug conjugates and as a PEG-based linker for PROTAC synthesis.

  • Fmoc-protected amine functionality for standard peptide chemistry
  • Terminal carboxylic acid for conjugation
  • High reported purity (≈99.8%)
  • Molecular weight 443.49 g/mol
  • Chemical formula C24H29NO7
  • Available in small-scale quantities, including 1 G

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.