Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,8,11-Trioxa-2-azatridecanoic acid, 13-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl)amino]-, 1,1-dimethylethyl ester | 2204246-03-5 | 97.2% | 548.59 | 5 MG
SDP

Catalog No. 5000403850
Encompass_Preferred

Thalidomide-NH-PEG3-NH-Boc is a chemical compound that appears as a light yellow to yellow oil. This product has been thoroughly tested and complies with the given specifications, ensuring its quality and suitability for research purposes.

  • Molecular formula: C26H36N4O9
  • Molecular weight: 548.59
  • Purity (LCMS): 97.20%
  • Storage for pure form: -20°C for 3 years; 4°C for 2 years
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-004-03850 Supplier Medchemexpress LLC Supplier No. HY440635MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-NH-PEG3-NH-Boc is a chemical compound that appears as a light yellow to yellow oil. This product has been thoroughly tested and complies with the given specifications, ensuring its quality and suitability for research purposes.

  • Molecular formula: C26H36N4O9
  • Molecular weight: 548.59
  • Purity (LCMS): 97.20%
  • Storage for pure form: -20°C for 3 years; 4°C for 2 years
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.