Learn More
Medchemexpress LLC 5,8,11-trioxa-2-azatridecanoic acid, 13-hydroxy-, 1,1-dimethylethyl ester | 106984-09-2 | 99.5% | 293.36 g·mol⁻¹ | C13H27NO6 | 25 G
SDP
Catalog No. 5000303017
Encompass_Preferred
Boc-NH-PEG4 is a PEG-based linker containing a Boc-protected amine, designed as a hydrophilic, flexible spacer for use in the synthesis of PROTACs and other bifunctional conjugates. It facilitates modular assembly and conjugation chemistry in medicinal chemistry workflows.
  • PEG spacer increases solubility and flexibility.
  • Boc-protected amine enables selective deprotection and downstream coupling.
  • High purity supports reproducible synthesis and analytical characterization.
  • Liquid form simplifies handling and weighing for small-scale reactions.
  • Suitable for assembling bifunctional degraders and other conjugates.
  • Stable under frozen storage for extended shelf life.
Catalog No. 50-003-03017 Supplier Medchemexpress LLC Supplier No. 25GHYW025896
Boc-NH-PEG4 is a PEG-based linker containing a Boc-protected amine, designed as a hydrophilic, flexible spacer for use in the synthesis of PROTACs and other bifunctional conjugates. It facilitates modular assembly and conjugation chemistry in medicinal chemistry workflows.
  • PEG spacer increases solubility and flexibility.
  • Boc-protected amine enables selective deprotection and downstream coupling.
  • High purity supports reproducible synthesis and analytical characterization.
  • Liquid form simplifies handling and weighing for small-scale reactions.
  • Suitable for assembling bifunctional degraders and other conjugates.
  • Stable under frozen storage for extended shelf life.