Learn More
Medchemexpress LLC 5,8,11-Trioxa-2-azatridecanoic acid, 13-hydroxy-, (1α,8α,9ẞ)-bicyclo[6.1.0]non-4-yn-9-ylmethyl ester | 2126749-77-5 | 99.6% | 369.45 g/mol | C19H31NO6 | 50 MG
SDP
Catalog No. 5000202514
Encompass_Preferred
BCN-PEG3-OH is a BCN-functionalized PEG linker intended for bioconjugation and antibody-drug conjugate (ADC) synthesis. The reagent contains a bicyclo[6.1.0]nonyne (BCN) group that reacts with azides via strain-promoted alkyne-azide cycloaddition (SPAAC), enabling copper-free click chemistry in aqueous conditions.
  • Non-cleavable four-unit PEG linker suitable for ADC construction.
  • Enables strain-promoted alkyne-azide cycloaddition with azide-functionalized molecules.
  • High reported purity for reliable conjugation performance.
  • Available in multiple pack sizes, including small research quantities.
Catalog No. 50-002-02514 Supplier Medchemexpress LLC Supplier No. HY13092550MG
BCN-PEG3-OH is a BCN-functionalized PEG linker intended for bioconjugation and antibody-drug conjugate (ADC) synthesis. The reagent contains a bicyclo[6.1.0]nonyne (BCN) group that reacts with azides via strain-promoted alkyne-azide cycloaddition (SPAAC), enabling copper-free click chemistry in aqueous conditions.
  • Non-cleavable four-unit PEG linker suitable for ADC construction.
  • Enables strain-promoted alkyne-azide cycloaddition with azide-functionalized molecules.
  • High reported purity for reliable conjugation performance.
  • Available in multiple pack sizes, including small research quantities.