Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,8,11-trioxa-2-azatridecanoic acid, 13-hydroxy-, (4E)-4-cycloocten-1-yl ester | 1955497-82-1 | 99.3% | 345.43 g·mol⁻¹ | C17H31NO6
SDP

Catalog No. 5000263032
Encompass_Preferred

TCO-PEG3-alcohol is a PEG-based PROTAC linker and click-chemistry reagent featuring a trans-cyclooctene (TCO) moiety and a terminal alcohol. It is supplied as a viscous, colorless to light-yellow liquid with high stated purity and recommended cold storage, and is intended for use in bioconjugation, PROTAC assembly, and other chemical biology applications.

  • Contains a trans-cyclooctene group for rapid iEDDA click reactions.
  • PEG3 spacer provides hydrophilicity and conformational flexibility.
  • Terminal alcohol enables additional derivatization and conjugation chemistry.
  • High purity suitable for synthetic and bioconjugation workflows.
  • Storage recommendations support long-term stability when kept cold.

Catalog No. 50-002-63032 Supplier Medchemexpress LLC Supplier No. HY1334765MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TCO-PEG3-alcohol is a PEG-based PROTAC linker and click-chemistry reagent featuring a trans-cyclooctene (TCO) moiety and a terminal alcohol. It is supplied as a viscous, colorless to light-yellow liquid with high stated purity and recommended cold storage, and is intended for use in bioconjugation, PROTAC assembly, and other chemical biology applications.

  • Contains a trans-cyclooctene group for rapid iEDDA click reactions.
  • PEG3 spacer provides hydrophilicity and conformational flexibility.
  • Terminal alcohol enables additional derivatization and conjugation chemistry.
  • High purity suitable for synthetic and bioconjugation workflows.
  • Storage recommendations support long-term stability when kept cold.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.