Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,8,11-trioxa-2-azatridecanoic acid, 13-hydroxy-, bicyclo[6.1.0]non-4-yn-9-ylmethyl ester | 2126749-77-5 | 99.6% | 369.45 | C19H31NO6 | 5 MG
SDP

Catalog No. 5000203923
Encompass_Preferred

BCN-PEG3-OH is a BCN-functionalized PEG linker with a terminal hydroxyl group used for bioorthogonal click chemistry to attach payloads in bioconjugation and antibody-drug conjugate synthesis.

  • enables strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • contains a three-unit polyethylene glycol spacer and a BCN reactive group.
  • high purity (99.6% by LCMS) and molecular weight 369.45 g/mol.
  • chemical formula C19H31NO6, cas 2126749-77-5.
  • store under nitrogen at -20°C; in solvent, -80°C for up to 6 months, -20°C for up to 1 month.

Catalog No. 50-002-03923 Supplier Medchemexpress LLC Supplier No. HY1309255MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BCN-PEG3-OH is a BCN-functionalized PEG linker with a terminal hydroxyl group used for bioorthogonal click chemistry to attach payloads in bioconjugation and antibody-drug conjugate synthesis.

  • enables strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • contains a three-unit polyethylene glycol spacer and a BCN reactive group.
  • high purity (99.6% by LCMS) and molecular weight 369.45 g/mol.
  • chemical formula C19H31NO6, cas 2126749-77-5.
  • store under nitrogen at -20°C; in solvent, -80°C for up to 6 months, -20°C for up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.