Learn More
Grainger Greenfield Threading Straight Flute Hand Tap Set, 5/8"-18 UNF, H3/3B, 4 Flute, HSS, 3-Piece Set
Catalog No. 502756639
Encompass_Preferred
This three-piece hand tap set is designed for precision thread cutting in a variety of materials including steel, stainless steel, cast iron, and non-ferrous metals. The set features high-speed steel construction with a bright, uncoated finish, ensuring durability and consistent performance in demanding threading applications.
  • Includes one each of taper, plug, and bottoming taps
  • Features four straight flutes for efficient chip removal
  • Designed to 5/8 in.-18 UNF thread size specifications
  • H3/3B class of fit and tolerance
  • Right-hand cutting direction
  • Compliant with USCTI 302 standards
Catalog No. 50-275-6639 Supplier Grainger Supplier No. 15J784
This three-piece hand tap set is designed for precision thread cutting in a variety of materials including steel, stainless steel, cast iron, and non-ferrous metals. The set features high-speed steel construction with a bright, uncoated finish, ensuring durability and consistent performance in demanding threading applications.
  • Includes one each of taper, plug, and bottoming taps
  • Features four straight flutes for efficient chip removal
  • Designed to 5/8 in.-18 UNF thread size specifications
  • H3/3B class of fit and tolerance
  • Right-hand cutting direction
  • Compliant with USCTI 302 standards