Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,8-Dibromoisoquinoline, 98%, Thermo Scientific™

Catalog No. AAH6303403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6303403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6303403 Supplier Thermo Scientific Chemicals Supplier No. H6303403

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81045-39-8
Molecular Formula C9H5Br2N
Molecular Weight (g/mol) 286.954
MDL Number MFCD03109888
InChI Key KNBALCRXZUTMBP-UHFFFAOYSA-N
PubChem CID 271281
IUPAC Name 5,8-dibromoisoquinoline
SMILES C1=CC(=C2C=NC=CC2=C1Br)Br

Specifications

Quantity 1 g
Synonym 5,8-dibromo-isoquinoline, isoquinoline, 5,8-dibromo, acmc-209pkl, 5,8-dibromo isoquinoline, isoquinoline,5,8-dibromo, 5,8-bis bromanyl isoquinoline
Formula Weight 286.95
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5,8-Dibromoisoquinoline
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.