Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,8-dihydroxy-3-methyl-9H-benzo[f]benzofuran-4-one | 150045-18-4 | MFCD00923100 | 99.5% | 230.22 g/mol | C13H10O4 | 1 ML
SDP

Catalog No. 5000214441
Encompass_Preferred

MS-444 is a small-molecule inhibitor that targets smooth muscle myosin light chain kinase (MLCK) and the RNA-binding protein HuR (ELAVL1). It inhibits purified MLCK with an IC50 of 10 μM and is supplied as a 10 mM solution in DMSO.

  • Inhibits MLCK with IC50 of 10 μM.
  • Also inhibits RNA-binding protein HuR (ELAVL1).
  • Supplied as a 10 mM solution in DMSO, 1 mL volume.
  • High purity (99.45%).
  • Molecular weight 230.22 g/mol; formula C13H10O4.
  • Recommended storage: -80°C for long-term storage of solutions; protect from light.

Catalog No. 50-002-14441 Supplier Medchemexpress LLC Supplier No. HY10068510MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MS-444 is a small-molecule inhibitor that targets smooth muscle myosin light chain kinase (MLCK) and the RNA-binding protein HuR (ELAVL1). It inhibits purified MLCK with an IC50 of 10 μM and is supplied as a 10 mM solution in DMSO.

  • Inhibits MLCK with IC50 of 10 μM.
  • Also inhibits RNA-binding protein HuR (ELAVL1).
  • Supplied as a 10 mM solution in DMSO, 1 mL volume.
  • High purity (99.45%).
  • Molecular weight 230.22 g/mol; formula C13H10O4.
  • Recommended storage: -80°C for long-term storage of solutions; protect from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.