Learn More
Sigma Aldrich 5,8-Dimethyldibenzo[b,j][1,10]phenanthroline-6,7-diol
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Ethyl 1,3-dithiane-2-carboxylate is an α-keto acid equivalent and bulky equivalent of acetate
- It participates in syn-selective aldol reactions
- It can be prepared from the reaction of ethyl diethoxyacetate and 1,3-propanedithiol in the presence of bf3/et2o
- Asymmetric oxidation of ethyl 1,3-dithiane-2-carboxylate by modena protocol has been reported to afford trans bis-sulfoxide in 60% yield
- Carbanion from ethyl 1,3-dithiane-2-carboxylate may be employed for the preparation of α-keto esters
Specifications
Specifications
| Molecular Formula | C22H16N2O2 |
| Linear Formula | C22H16N2O2 |
| MDL Number | MFCD07626356 |
| Quantity | 5 mL |
| Molecular Weight (g/mol) | 340.37 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.