Learn More
Medchemexpress LLC 5,8-dioxa-2,11-diazatetradecanoic acid, 14-(2,5-dihydro-2,5-dioxo-1H-pyrrol-1-yl)-12-oxo... | 2675364-08-4 | 98.1% | 475.53 | C24H33N3O7 | 5 MG
SDP
Catalog No. 5000204461
Encompass_Preferred
BCN-endo-PEG2-maleimide is a bioconjugation reagent combining an endo-BCN strained alkyne and a maleimide thiol-reactive group linked by a PEG2 spacer. It is intended for antibody-drug conjugate (ADC) linker synthesis, click chemistry (SPAAC), and thiol conjugation in bioconjugation workflows.
  • Provides strain-promoted azide-alkyne cycloaddition reactivity (endo-BCN)
  • Contains a maleimide group for thiol-selective conjugation
  • Short PEG2 spacer improves solubility and flexibility
  • High purity suitable for research (purity 98.06%)
  • Available in small pack sizes for conjugation optimization
Catalog No. 50-002-04461 Supplier Medchemexpress LLC Supplier No. HY1563115MG
BCN-endo-PEG2-maleimide is a bioconjugation reagent combining an endo-BCN strained alkyne and a maleimide thiol-reactive group linked by a PEG2 spacer. It is intended for antibody-drug conjugate (ADC) linker synthesis, click chemistry (SPAAC), and thiol conjugation in bioconjugation workflows.
  • Provides strain-promoted azide-alkyne cycloaddition reactivity (endo-BCN)
  • Contains a maleimide group for thiol-selective conjugation
  • Short PEG2 spacer improves solubility and flexibility
  • High purity suitable for research (purity 98.06%)
  • Available in small pack sizes for conjugation optimization