Learn More
Medchemexpress LLC 5,8-dioxa-2,11-diazatetradecanoic acid 14-(2,5-dihydro-2,5-dioxo-1H-pyrrol-1-yl) methyl ester | 2675364-08-4 | 98.1% | 475.53 | C24H33N3O7 | 10 MG
SDP
Catalog No. 5000204430
Encompass_Preferred
BCN-endo-PEG2-maleimide is an ADC linker that combines an endo-bicyclononyne (BCN) reactive group, a PEG2 spacer, and a maleimide thiol-reactive moiety for use in click chemistry and thiol conjugation workflows.
  • High purity suitable for conjugation (98.1%).
  • Provides a BCN moiety for strain-promoted azide-alkyne cycloaddition.
  • Maleimide group enables selective thiol coupling.
  • PEG2 spacer improves aqueous solubility and reduces steric hindrance.
  • Molecular weight 475.53 g/mol; formula C24H33N3O7.
  • Available in small research quantities (for example, 10 mg).
Catalog No. 50-002-04430 Supplier Medchemexpress LLC Supplier No. HY15631110MG
BCN-endo-PEG2-maleimide is an ADC linker that combines an endo-bicyclononyne (BCN) reactive group, a PEG2 spacer, and a maleimide thiol-reactive moiety for use in click chemistry and thiol conjugation workflows.
  • High purity suitable for conjugation (98.1%).
  • Provides a BCN moiety for strain-promoted azide-alkyne cycloaddition.
  • Maleimide group enables selective thiol coupling.
  • PEG2 spacer improves aqueous solubility and reduces steric hindrance.
  • Molecular weight 475.53 g/mol; formula C24H33N3O7.
  • Available in small research quantities (for example, 10 mg).