Learn More
Medchemexpress LLC 5,8-dioxa-2,11-diazatetradecanoic acid bicyclo[6.1.0]non-4-yn-9-ylmethyl ester | 2675364-08-4 | 98.1% | 475.53 g/mol | C24H33N3O7 | 50 MG
SDP
Catalog No. 5000204498
Encompass_Preferred
BCN-endo-PEG2-maleimide is an ADC linker containing PEG spacer units, an endo-BCN strained cyclooctyne moiety, and a maleimide reactive group used for bioconjugation and click-chemistry applications. It is employed in antibody-drug conjugate research to attach payloads to azide- or thiol-containing partners, and is reported to be an oil with high purity.
  • Used as an ADC linker and for click-chemistry conjugations
  • Contains endo-BCN for rapid bioorthogonal reaction with azides
  • Features a maleimide group for thiol-reactive conjugation
  • Reported purity approximately 98.1% and appears as an oil
  • Store protected from light; in solvent: -80°C (6 months), -20°C (1 month)
Catalog No. 50-002-04498 Supplier Medchemexpress LLC Supplier No. HY15631150MG
BCN-endo-PEG2-maleimide is an ADC linker containing PEG spacer units, an endo-BCN strained cyclooctyne moiety, and a maleimide reactive group used for bioconjugation and click-chemistry applications. It is employed in antibody-drug conjugate research to attach payloads to azide- or thiol-containing partners, and is reported to be an oil with high purity.
  • Used as an ADC linker and for click-chemistry conjugations
  • Contains endo-BCN for rapid bioorthogonal reaction with azides
  • Features a maleimide group for thiol-reactive conjugation
  • Reported purity approximately 98.1% and appears as an oil
  • Store protected from light; in solvent: -80°C (6 months), -20°C (1 month)