Learn More
Medchemexpress LLC 5,8R-dihydro-N-[(1S)-2,2,2-trifluoro-1-methylethyl]-8-[4-(trifluoromethyl)phenyl]-1,7-naphthyridine-7 | 1470018-52-0 | 99.9% | 417.35 | C19H17F6N3O | 10 MG
SDP
Catalog No. 5000243958
Encompass_Preferred
AMG2850 is a potent, orally bioavailable, and selective TRPM8 antagonist designed for preclinical research of TRP channels and pain modalities. It is supplied as a high-purity solid and as a DMSO solution for in vitro and in vivo studies, with recommended storage conditions for long-term stability.
  • Potent, orally bioavailable TRPM8 antagonist.
  • High purity (99.9%).
  • Available as powder and 10 mM solution in DMSO.
  • High solubility in DMSO (100 mg/mL) and formulatable for in vivo dosing.
  • Stable when stored at recommended temperatures for long-term use.
  • Molecular weight 417.35 and chemical formula C19H17F6N3O.
Catalog No. 50-002-43958 Supplier Medchemexpress LLC Supplier No. HY10405910MG
Add to Cart
Add to Cart
AMG2850 is a potent, orally bioavailable, and selective TRPM8 antagonist designed for preclinical research of TRP channels and pain modalities. It is supplied as a high-purity solid and as a DMSO solution for in vitro and in vivo studies, with recommended storage conditions for long-term stability.
  • Potent, orally bioavailable TRPM8 antagonist.
  • High purity (99.9%).
  • Available as powder and 10 mM solution in DMSO.
  • High solubility in DMSO (100 mg/mL) and formulatable for in vivo dosing.
  • Stable when stored at recommended temperatures for long-term use.
  • Molecular weight 417.35 and chemical formula C19H17F6N3O.