Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,9-Undecadien-2-one, 6,10-dimethyl- | 689-67-8 | 100 G
SDP

Catalog No. 5000417409
Encompass_Preferred

(E/Z)-Geranylacetone is an organic compound commonly used in fragrances and flavors, imparting a fresh aromatic smell to various products. It also serves as a versatile biochemical reagent for life science research. Its applications extend to cosmetics, certain foods, and pharmaceuticals.

  • Molecular weight of 194.31
  • Chemical formula C13H22O
  • Appearance is a colorless to light yellow liquid
  • Purity of 97.0%
  • Used in perfumes, soaps, and cosmetics
  • Applied in foods, such as candies and chewing gum
  • Found in herbal remedies and pharmaceuticals
  • Functions as a biochemical reagent
  • Originally sourced from plants such as Solanaceae Nicotiana tabacum L.

Catalog No. 50-004-17409 Supplier Medchemexpress LLC Supplier No. HYN8446100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E/Z)-Geranylacetone is an organic compound commonly used in fragrances and flavors, imparting a fresh aromatic smell to various products. It also serves as a versatile biochemical reagent for life science research. Its applications extend to cosmetics, certain foods, and pharmaceuticals.

  • Molecular weight of 194.31
  • Chemical formula C13H22O
  • Appearance is a colorless to light yellow liquid
  • Purity of 97.0%
  • Used in perfumes, soaps, and cosmetics
  • Applied in foods, such as candies and chewing gum
  • Found in herbal remedies and pharmaceuticals
  • Functions as a biochemical reagent
  • Originally sourced from plants such as Solanaceae Nicotiana tabacum L.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.