Learn More
Medchemexpress LLC HY-131296 5mg , 5-A-RU-PABC-Val-Cit-Fmoc CAS:2677841-58-4 Purity:>98%
SDP
Catalog No. 502259487
Encompass_Preferred
HY-131296 5mg 5-A-RU-PABC-Val-Cit-Fmoc CAS: 2677841-58-4 5-A-RU-PABC-Val-Cit-Fmoc is the prodrug of 5-A-RU. 5-A-RU, a precursor of bacterial Riboflavin, is a mucosal-associated invariant T (MAIT) cells activator. 5-A-RU forms potent MAIT-activating antigens via non-enzymatic reactions with small molecules, such as glyoxal and methylglyoxal, which are derived from other metabolic pathways[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9487 Supplier Medchemexpress LLC Supplier No. HY1312965MG
HY-131296 5mg 5-A-RU-PABC-Val-Cit-Fmoc CAS: 2677841-58-4 5-A-RU-PABC-Val-Cit-Fmoc is the prodrug of 5-A-RU. 5-A-RU, a precursor of bacterial Riboflavin, is a mucosal-associated invariant T (MAIT) cells activator. 5-A-RU forms potent MAIT-activating antigens via non-enzymatic reactions with small molecules, such as glyoxal and methylglyoxal, which are derived from other metabolic pathways[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.